About this item
IntelliCAL PHC725 is a digital, combination glass-body probe with a refillable KCl reference and built-in temperature sensor.
- High performance Red Rod technology now available for the digital HQD platform
- Red Rod technology provides fast, accurate and reproducible measurements
- Red Rod reliability when measuring challenging samples over wider temperature ranges
- IntelliCAL™ digital probes provide ultimate traceability in measurement history
IntelliCAL™ digital probes can be moved between meters without the need to re-calibrate or re-enter measurement settings. It has an annular ring junction and is ideal for measuring pH in a variety of applications, including laboratory measurement of drinking water and low ionic strength (LIS) solutions.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10146-000
- Cable length:
- 1 m
- Junction:
- Annular ring
- Temperature range:
- −10...+100 °C
- Range:
- 0 - 14 pH
- Accuracy:
- ±0.01 pH
- IP rating:
- IP 65
- Filling solution:
- Sat. KCl