Anti-RAP2C Rabbit Polyclonal Antibody
Catalog # 89514-190
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rap-2C
- Antigen symbol:RAP2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:57826
- Amino acid number:14 to 42
- Storage buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular weight:20.745 kDa
- Sequence:IEKYDPTIEDFYRKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping temperature:4 °C
- Immunogen:This RAP2C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 14-42 amino acids from the N-terminal region of human RAP2C.
- Purification:Protein A & Peptide affinity purification
- Cat. no.:89514-190
- Supplier No.:AP19868A
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: RAP2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse