Calcium Standard Solution, 1000 mg/L as CaCO₃, 1 L
Catalog # CA121-53
Supplier: Hach
CAS Number:
Specifications
- Concentration:1000 mg/L as CaCO₃
- Description:Calcium
- Size:1 L
- Cat. no.:CA121-53
- Supplier No.:121-53
Specifications
About this item
For accuracy checks in calcium hardness determinations.