Anti-IL-27 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B032F6]
Supplier: Biolegend
Certificates
About this item
PE anti-human IL-27/IL-35 EBI3 (monomer, dimer, heterodimer) [B032F6]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IL-27
Clonality: Monoclonal
Clone: B032F6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Specifications
- Catalog No:
- CA10760-796
- CA10760-794
- Antigen symbol:
- IL-27
- IL-27
- Antigen synonyms:
- IL-27B|Epstein Barr virus induced gene 3|EBI3|IL-35 subunit
- IL-27B|Epstein Barr virus induced gene 3|EBI3|IL-35 subunit
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- B032F6
- B032F6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 10148
- 10148
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Immunogen:
- Purified human IL-27
- Purified human IL-27
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together