Order Entry
Canada
ContactUsLinkComponent
Anti-CD45.1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: A20]
Anti-CD45.1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: A20]
Supplier:  Biolegend
Certificates
Anti-CD45.1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: A20]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • CA10756-350
  • CA110737-BL
  • Antigen symbol:
  • CD45.1
  • CD45.1
  • Antigen synonyms:
  • PTPRC|T200|LCA|Ly-5.1
  • PTPRC|T200|LCA|Ly-5.1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Brilliant Violet® 605
  • Brilliant Violet® 605
  • Clone:
  • A20
  • A20
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • SJL mouse thymocytes and splenocytes
  • SJL mouse thymocytes and splenocytes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 605™ and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 605™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 605™ and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • Storage temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together