ContactUsLinkComponent
Clean-Cut Tubing Cutter, Restek
Clean-Cut Tubing Cutter, Restek
Catalog # 10059-354
Supplier:  Restek
undefined
Clean-Cut Tubing Cutter, Restek
Catalog # 10059-354
Supplier:  Restek
Supplier Number:  25069
Specifications
  • Description:
    Polymeric Tubing Cutter
  • Cat. no.:
    10059-354
  • Cutting Capacity:
    ⅛" Ext.Ø
  • Supplier No.:
    25069
Specifications
About this item
Clean-Cut tubing cutter uses on PEEK, PTFE, Tefzel®, and other polymeric tubing.
It is burr-free, with perpendicular cuts that will not distort the tubing OD or close the ID.
Ideal to use with Agilent HPLC systems.