Rtx®-VMS Columns (Fused Silica), Restek
: Restek
Rtx®-VMS columns offer lower bleed, better selectivity, and overall faster analysis for separating volatile organic compounds, such as those listed in U.S.
- Application-specific columns for analyzing volatile organic pollutants by GC-MS including methods TO-15, TMS, and EPA 8260
- Complete separation of U.S. EPA method 8260 compounds in less than 10 minutes
- Stable to 260 °C
- No known equivalent phases
- Proprietary Crossbond® phase
The Rtx®-VMS stationary phase is a highly stable polymer that provides outstanding analysis of volatile compounds, in combination with sensitive ion traps and Agilent 5973 mass spectrometers. 0.18 and 0.25 mm Int.Ø. columns allow sample splitting at the injection port, eliminating the added expense and maintenance of a jet separator. A 0.45 or 0.53 mm Int.Ø. column can be directly connected to the purge-and-trap transfer line in a system equipped with a jet separator.
EPA method 8260B.
- Catalog No:
- 10850-524
- 10850-568
- 10850-526
- 10850-528
- 10850-530
- 10850-532
- 10850-690
- 10850-692
- 10850-522
- 10850-566
- Description:
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Rtx-VMS
- Temperature limit:
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
- -40 to 240/260 °C
Frequently Bought Together