MXT®-Volatiles Columns (Siltek®-treated stainless steel), Restek
Supplier: Restek
Certificates
About this item
MXT®-Volatiles columns were the first columns designed specifically for analyses of the 34 volatile organic pollutants listed in U.S.
- Application-specific columns for volatile organic pollutants
- Temperature range: −20 to 320 °C
- 4.5" standard coil diameter
- Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase
With these columns, you can quantify all compounds listed in these methods, whether you use a mass spectrometer or a PID in tandem with an ELCD. The diphenyl/dimethyl polysiloxane based MXT®-Volatiles stationary phase provides low bleed and thermal stability to 320 °C.
EPA methods 601, 602, and 624.
Specifications
- Catalog No:
- 10851-488
- 10851-490
- 10851-492
- 10851-494
- Description:
- MXT-Volatiles
- MXT-Volatiles
- MXT-Volatiles
- MXT-Volatiles
- Temperature limit:
- -20 to 280/320 °C
- -20 to 280/320 °C
- -20 to 280/320 °C
- -20 to 280/320 °C