Order Entry
Canada
ContactUsLinkComponent
Downing Cartilage Knife, OR Grade, Sklar
Downing Cartilage Knife, OR Grade, Sklar
Catalog # 10196-878
Supplier:  Sklar
undefined
Downing Cartilage Knife, OR Grade, Sklar
Catalog # 10196-878
Supplier:  Sklar
Supplier Number:  47-3415

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Downing Cartilage Knife, Slightly Curved, Concave Edge with Guards
  • Length:
    25.4 cm (10")
  • Material:
    Stainless steel
  • Cat. no.:
    10196-878
  • Supplier No.:
    47-3415

Specifications

About this item

Downing Cartilage Knives are constructed of stainless steel

Industry standard.