Orders LinkContactUsLinkComponent
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Supplier:   Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Supplier:  Corning
Certificates
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
3 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.
 : Triple 0.1 µm sterilized.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA45000-300
  • CA45000-302
  • CA45000-504
  • Storage temperature:
  • 2...8 °C
  • 2...8 °C
  • 2...8 °C
Specifications
Product Family Options
Product Information
  • Media Format
    Size
    Availability
    Price
  • CA45000-300
    Minimum Essential Medium (MEM) Alpha Medium with Earle's salts, ribonucleosides, deoxyribonucleosides, and L-glutamine
    Minimum Essential Medium (MEM) Alpha Medium with Earle's salts, ribonucleosides, deoxyribonucleosides, and L-glutamine
    Liquid
    500 ml
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $296.00
    $296.00
    Pack of 6 (500ml)
       $296.00
    Supplier Number: 10-022-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-300
    Storage temperature
    2...8 °C
  • CA45000-302
    Minimum Essential Medium (MEM) Alpha Medium with Earle's salts without ribonucleosides, deoxyribonucleosides, and no L-glutamine
    Minimum Essential Medium (MEM) Alpha Medium with Earle's salts without ribonucleosides, deoxyribonucleosides, and no L-glutamine
    Liquid
    500 ml
    Log in for availability
    Log in for availability
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $235.00
    $235.00
    Pack of 6 (500ml)
       $235.00
    Supplier Number: 15-012-CV
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-302
    Storage temperature
    2...8 °C
  • CA45000-504
    Minimum Essential Medium (MEM) Alpha Medium, Powder with Earle's salts and L-glutamine without ribonucleosides, deoxyribonucleosides, and sodium bicarbonate
    Minimum Essential Medium (MEM) Alpha Medium, Powder with Earle's salts and L-glutamine without ribonucleosides, deoxyribonucleosides, and sodium bicarbonate
    Powder
    50 L
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $121.00
    $121.00
    Each (50l)
       $121.00
    Supplier Number: 50-012-PC
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA45000-504
    Storage temperature
    2...8 °C