Order Entry
Canada
ContactUsLinkComponent
Reiner Plaster Knife, OR Grade, Sklar
Reiner Plaster Knife, OR Grade, Sklar
Catalog # 10194-974
Supplier:  Sklar
undefined
Reiner Plaster Knife, OR Grade, Sklar
Catalog # 10194-974
Supplier:  Sklar
Supplier Number:  11-1550

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Reiner Plaster Knife with Metal Handle
  • Length:
    17.8 cm (7")
  • Material:
    Stainless steel
  • Cat. no.:
    10194-974
  • Supplier No.:
    11-1550

Specifications

About this item

Reiner Plaster Knives are constructed of stainless steel

This knife features a curved blade, with a 11/2" blade. Industry standard.