Order Entry
Canada
Orders LinkContactUsLinkComponent
Anti-PLXDC1 Rabbit Polyclonal Antibody

Anti-PLXDC1 Rabbit Polyclonal Antibody

400 µL
Catalog # 89514-762
Supplier:   Abgent
Supplier #:  AP19440B
Anti-PLXDC1 Rabbit Polyclonal Antibody
Catalog # 89514-762
Supplier:  Abgent
Anti-PLXDC1 Rabbit Polyclonal Antibody

400 µL

Price
$1,057.06
Log in for availability
Each (1mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Western Blot: 1:1000

Type: Primary
Antigen: PLXDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Documents

Specifications
  • Size:
    400 µL
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Protein A & Peptide affinity purification
  • Antibody type:
    Primary
  • Epitope:
    C-Terminal
  • Amino acid number:
    466 to 494
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Host:
    Rabbit
  • Sequence:
    YAEVEPSGHEKEGFC
  • Concentration:
    0.4mg/ml
  • Molecular weight:
    55.76 kDa
  • Shipping temperature:
    4 °C
  • Antigen name:
    Plexin Domain-Containing Protein 1
  • Cat. No.:
    89514-762
  • Reactivity:
    Human
  • Gene ID:
    57125
  • Antigen synonyms:
    TEM3|TEM7|Plexin domain-containing protein 1|Tumor endothelial marker 3
  • Immunogen:
    This PLXDC1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 466-494 amino acids from the C-terminal region of human PLXDC1.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    PLXDC1
  • Storage buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.