Anti-a-GalCer:CD1d complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
PE anti-mouse α-GalCer:CD1d complex [L363]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: a-GalCer:CD1d complex
Clonality: Monoclonal
Clone: L363
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
- Catalog No:
- CA140505-BL
- CA140506-BL
- Antigen symbol:
- a-GalCer:CD1d complex
- a-GalCer:CD1d complex
- Antigen synonyms:
- Ly-38|CD1.1|AI747460|CD1D1|Cd1d|Cd1a
- Ly-38|CD1.1|AI747460|CD1D1|Cd1d|Cd1a
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- L363
- L363
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 12479
- 12479
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Immunogen:
- α-GalCer:mCD1d1 complexes coupled to Mycobacterium tuberculosis PPD
- α-GalCer:mCD1d1 complexes coupled to Mycobacterium tuberculosis PPD
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together