SGE HT8-PCB GC Capillary Columns
Supplier: Trajan Scientific and Medical
Certificates
About this item
The SGE HT8-PCB GC Capillary Column provides a great low-polarity option for GC analyses. Recommended for: PCB congener analyses, nitro-substituted aromatics, polynuclear aromatic hydrocarbons, pesticides/herbicides.
- Preferred column for polychlorinated biphenyl (PCB) compounds
- Separates PCBs on ortho ring substitution as well as boiling point
- Ideal for environmental analysis
- Unique high temperature phase suited for the analysis of persistent organic pollutants (POPs)
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.
Specifications
- Catalog No:
- 10811-186
- 14226-014
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Temperature limit:
- 350 °C
- 370 °C
Specifications
Related Information
TDS HT8 The Perfect PCB Column.pdf
Datasheet
Trajan Scientific GC Consumables Catalog
Product Literature
Frequently Bought Together