Ultra Cyano Columns, Restek
Supplier: Restek
Certificates
About this item
The Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired.
- Particle size and pore Size: 3 or 5 µm, spherical and 100 Å
- Carbon load: 8%
- End-cap: yes
- pH range and temperature limit:2.5 to 8 and 80 °C
- USP phase code: L10
- Phase category: cyano
- Ligand type: cyanopropylsilane
It can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes. It is an excellent choice for the analysis of protonated bases.
Specifications
- Catalog No:
- 10058-340
- 10838-048
- 10838-040
- 10837-532
- 10837-530
- 10839-262
- 10838-056
- 10840-582
- 10837-526
- 10837-524
- 10837-520
- 10837-518
- 10837-544
- 10837-542
- 10838-026
- 10838-064
- 10058-336
- 10058-338
- 10837-548
- 10058-334
- Description:
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- USP code:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
Specifications
Frequently Bought Together