Order Entry
Canada
ContactUsLinkComponent
Concentric Ring Round Water Bath, Boekel Scientific
Concentric Ring Round Water Bath, Boekel Scientific
Catalog # 13540-044
Supplier:  Boekel
undefined
Concentric Ring Round Water Bath, Boekel Scientific
Catalog # 13540-044
Supplier:  Boekel
Supplier Number:  14458

Specifications

  • Capacity:
    1.75 L
  • Description:
    Round Water Bath with Seven Concentric Rings
  • Environmentally Preferable:
  • Cat. no.:
    13540-044
  • Supplier No.:
    14458
  • Ø×H:
    8.0×2.75"

Specifications

About this item

The Round Water Bath is made from brushed stainless steel for corrosion resistance and is supplied with seven concentric rings

The Boekel Scientific Concentric Ring Water Bath accommodates many vessel and tube sizes with the smallest opening of 25mm and largest opening of 203mm.

This water bath provides gentle heating of flasks and bottles while minimizing water loss through evaporation.

Ordering information: This unit includes 7 concentric rings.