Empty Centrifuge Tubes, FEP, Restek
Catalog # 10015-354
Supplier: Restek
Specifications
- Description:Empty 50 ml Centrifuge Tube, FEP
- Cat. no.:10015-354
- Supplier No.:23997
Specifications
About this item
Sturdy, easy-to-clean tubes last indefinitely under normal usage conditions and cost 50% less than traditional centrifuge tubes.
- Volume of 50 ml
- Sturdy
- Easy to clean
- FEP fabrication
- SPE accessory