ContactUsLinkComponent
Retaining Ring Plier for ASE® 100/200/300 Systems, Restek
Retaining Ring Plier for ASE® 100/200/300 Systems, Restek
Catalog # 10015-294
Supplier:  Restek
undefined
Retaining Ring Plier for ASE® 100/200/300 Systems, Restek
Catalog # 10015-294
Supplier:  Restek
Supplier Number:  26185
Specifications
  • Description:
    Retaining Ring Plier
  • Cat. no.:
    10015-294
  • Supplier No.:
    26185
Specifications
About this item
Durable, ergonomically designed pliers are effective at removing retaining rings, both internal and external.
  • Can be used for internal or external retaining rings
  • Works to remove retaining ring in all ASE® cell caps
For use with ASE® 100/200/300 systems.