Order Entry
Canada
ContactUsLinkComponent
 
Choice® MicroFiber String Mop, Berkshire
  89496-572
 :  Berkshire
Small Business Enterprise
undefined
Choice® MicroFiber String Mop, Berkshire
  89496-572
 :  Berkshire
 :  CHMFM.24
Small Business Enterprise

 

  • Color:
    White with Blue Head- and Tailband
  • Material:
    Polyester 88%/nylon microfiber 12%
  • Packed:
    Individual
  • Cat. no.:
    89496-572
  • Supplier No.:
    CHMFM.24

 

 

Microfibers are tiny fibers split to produce ultra-fine fibers.

  • White 88% polyester, 12% nylon microfiber string mop with blue headband and tailband
  • Microfiber yarns attract and trap dirt and debris
  • Terry knit structure enhances absorbency to increase efficiency
  • Lot number assures traceability
  • Can be laundered and re-used
  • ISO Class 5–8
  • Cleanroom Class 100–100,000

Terry knit structure enhances absorbency to increase efficiency. Tubular knit construction with sewn head and tail bands provides added durability. Mop can be laundered and re-used. Works with most standard loop mop frames and bucket/wringer systems.

Applications include, floor cleaning in critical environments, applying of disinfectants and general spill pick-up.

When woven together these fibers create a surface area covered with millions of spaces between the fibers to trap moisture, dirt and debris. This technology allows for the dirt and liquids to be 'picked up' rather than being pushed around. Microfiber yarns attract and trap dirt and debris.

 : 24 mops per case.