Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Universal Pathogen DNA Kit, Omega Bio-Tek
Mag-Bind® Universal Pathogen DNA Kit, Omega Bio-Tek
Catalog # CA10821-238
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Universal Pathogen DNA Kit, Omega Bio-Tek
Catalog # CA10821-238
Supplier:  Omega Bio-Tek
Supplier Number:  M4029-01
CAS Number:  

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Viral DNA
    Bacterial DNA
  • Sample type:
    Stool
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    MagBind Universal Pathogen DNA Kit
  • Size:
    4 x 96 Preps
  • Description:
    MagBind Universal Pathogen DNA Kit
  • Cat. no.:
    CA10821-238
  • Supplier No.:
    M4029-01

Specifications

About this item

The MagBind Universal Pathogen 96 Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples

  • No organic extractions
  • Pure DNA for downstream applications such as qPCR and next generation sequencing (NGS)
  • Isolate DNA and viral RNA from a wide variety of sample types, including feces

The system allows for automation after sample lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen BioSprint, and other liquid handling instruments.

E-Z 96 Disruptor Plus plates are included with the kit for simultaneous homogenization and lysis of the sample. The unique buffer system does not require alcohol to bind nucleic acids allowing for isolation of high-quality DNA/RNA, free of PCR inhibitors. The entire procedure can be completed in less than 60 minutes for 96-samples.