- Cat. no.:89237-584
- Packaging:Flat
- Supplier No.:SSP120.0110WFI
Sterile SatPax® MicroSeal® WFI combines MicroSeal® 1200 ultrasonically sealed edge cleanroom laundered wipers composed of 100% continuous filament polyester knit fabric with a high saturation level of 70% IPA and 30% WFI water.
This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated with wetting a dry wiper. Each package is then gamma irradiated sterile to a 10⁻⁶ Sterility Assurance Level making the product ideal for cleaning critical sterile processing environments.
Pre-wetted with consistent 70% IPA/ 30% WFI (Water for Injection) to a high saturation level. Details: expiration date, radiation dosage, lot information, Meets cGMP requirements for traceability.WFI (Water for Injection) solution for critical applications requiring WFI.Designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required.