Orders LinkContactUsLinkComponent
Ward's® Plastic Slide Boxes
Ward's® Plastic Slide Boxes
Supplier:   Ward's Science
Ward's® Plastic Slide Boxes
Supplier:  Ward's Science
Certificates
Ward's® Plastic Slide Boxes
Ward's® Plastic Slide Boxes
2 Options Available Below
Product Details & Documents
The plastic slide box protects 1x3" slides and won't warp when temperature and humidity vary.
  • Plastic construction
  • Foam-lined bottom
  • Numbered insert for easy organization of slides
The 25-capacity and 100-capacity boxes have fully removable lid latches with a plastic clip. The 10-capacity boxes are hinged, with a snap-close mechanism.
Product Family Options
Product Information
  • Availability
    Price
  • 470151-632
    25-Capacity Slide Box
    25-Capacity Slide Box
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $7.65
    $7.65
    Log in for availability
    Case of 12
    Case of 12 - $82.35
    $82.35
    Each
       $7.65
    Case of 12
       $82.35
    Supplier Number: 481010-R
  • 470151-630
    100-Capacity Slide Box
    100-Capacity Slide Box
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $24.65
    $24.65
    Log in for availability
    Case of 6
    Case of 6 - $140.40
    $140.40
    Each
       $24.65
    Case of 6
       $140.40
    Supplier Number: 481030-R