ContactUsLinkComponent
Expandable EZ LockPlatform Rack and Pallet, Quantum Storage Systems
Expandable EZ LockPlatform Rack and Pallet, Quantum Storage Systems
Catalog # 10804-428
undefined
Expandable EZ LockPlatform Rack and Pallet, Quantum Storage Systems
Catalog # 10804-428
Supplier Number:  EL-12406
Specifications
  • Load Capacity:
    2500 lbs.
  • Description:
    Expandable EZ Lock Platform, Rack and Pallet
  • Cat. no.:
    10804-428
  • Supplier No.:
    EL-12406
Specifications
About this item
Alllows you to customize your storage space in seconds.
  • Manufactured of recylced high density polyethylene
One piece body design and structure allows for a 4 way forklift and hand truck entry, and meets GMA industry standards.
Temperature rated -30 to 200o C and offers 2500 lbs. capacity when connected. Plastic will not absorb any liquids or chemicals and is resistant to insect infestation. Sold in two pieces to be interlocked. Black color.
Interlocking design allows flexibility in building exact size pallet or platform.