To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:PRB-1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Size:1 Kit
- Cross adsorption:No
- Storage temperature:The APO-BrdU Kit is shipped in one container and consists of two packages. Upon arrival one should be stored at 2-8°C and the other at –20 °C.
- Cat. no.:10820-970
The fragmentation of genomic DNA by cellular nucleases during the later stages of apoptosis is also one of the most easily measured features of apoptotic cells. Nuclease activity generates DNA fragments ranging from ~300 bp to 50 bp in length, resulting in a typical DNA ‘laddering’ appearance when analyzed by agarose gel electrophoresis. These fragments have exposed 3’-hydroxyl (OH) ends which can be labeled with bromolated deoxyuridine triphosphates (Br-dUTP). An enzyme, terminal deoxynucleotidyl transferase (TdT), is used to catalyze the template-independent addition of Br-dUTP to the 3’-OH ends of double or single stranded DNA. This method is often called TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) or end labeling. Sites where the Br-dUTP is incorporated can then be detected with an antibody specific to BrdU. With the APO-BrdUTM Kit cells are first labeled with Br-dUTP, and then sites of incorporation are detected through staining with a FITC anti-BrdU antibody. Samples can then be analyzed via flow cytometry. Samples that are apoptotic will stain brightly with the anti-BrdU antibody due to the substantial number of exposed 3’-OH sites, while cells that are non-apoptotic will not have incorporated significant amounts of Br-dUTP and will stain dimly. The APO-BrdU Kit is shipped in one container and consists of two packages. Upon arrival one should be stored at 2-8°C and the other at -20°C.
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: PRB-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: 0