If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Length:121 mm (4¾")
- Material:Tungsten carbide/stainless steel
- Tip Configuration:7 x 7 Teeth
- Cat. no.:89025-094
- Jaw Type:Cross-serrated
- Tip Style:Straight
- Supplier No.:51-1448
Specifications
About this item
LiteGrip™ Tungsten Carbide Adson Tissue Forceps are for holding and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
LiteGrip™ instruments feature a lightweight, fenestrated handle. They are useful during lengthy procedures when fatigue may arise. The finger holes on the handle also allow grip adjustments as needed by the surgeon.