Order Entry
Canada
ContactUsLinkComponent
Silicon, AAS Standard Solution, Specpure®, Si 1000 µg/ml, Thermo Scientific
Silicon, AAS Standard Solution, Specpure®, Si 1000 µg/ml, Thermo Scientific
Catalog # AA47277-AE
CAS Number:  
Silicon, AAS Standard Solution, Specpure®, Si 1000 µg/ml, Thermo Scientific
Catalog # AA47277-AE
Supplier Number:  047277-AE
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O/tr. HF
  • Size:
    100 ml
  • MDL No.:
    MFCD00085311
  • Cat. no.:
    AA47277-AE
  • Element:
    Silicon
  • Supplier No.:
    47277-AE

Specifications

About this item

Molecular Formula: (NH4)2SiF6 in H2O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00085311