Terbium, Plasma Standard Solution, Specpure®, Tb 10 µg/ml, Thermo Scientific
AA45266-AE
:
- Concentration:10 µg/ml
- Matrix:2% HNO₃
- Size:100 ml
- MDL No.:MFCD00011256
- Cat. no.:AA45266-AE
- Element:Terbium
- Supplier No.:45266-AE
Molecular Formula: Tb4O7 in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011256