ORP test solution, 200-275 mV, Thermo Scientific
Fournisseur: Thermo Scientific Chemicals
Certificats
À propos de cet article
Physical Form: Liquid
MDL No.: MFCD01867801
Test solution for ORP electrodes
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- MDL No.:MFCD01867801
Spécifications
Informations connexes
Certificats
Recherche par numéro de lotAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Frequently Bought Together