ContactUsLinkComponent
Sklar Black™ Thumb Tissue Forceps, OR Grade, Sklar
  10197-122
 :  Sklar
undefined
Sklar Black™ Thumb Tissue Forceps, OR Grade, Sklar
  10197-122
 :  Sklar
 :  90-1023
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Sklar Black™ Thumb Tissue Forceps
  • Length:
    6"
  • Material:
    Stainless Steel with Reduced Reflectivity Polymer Coating
  • Tip Configuration:
    1 x 2 Teeth
  • Cat. no.:
    10197-122
  • Supplier No.:
    90-1023
Sklar Black™ Thumb Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in laser procedures
These instruments have spring tension shanks that ultimately determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects. This product is straight with 1x2 teeth, and a length of 6 inches.