Order Entry
Canada
ContactUsLinkComponent
Microtiter Tubes, Porex Bio Products Group
Microtiter Tubes, Porex Bio Products Group
Catalog # 46300-494
Supplier:  Thermo Scientific
undefined
Microtiter Tubes, Porex Bio Products Group
Catalog # 46300-494
Supplier:  Thermo Scientific
Supplier Number:  845

Specifications

  • Sterility:
    Non sterile
  • Capacity:
    1.2 ml
  • Color:
    Transparent
  • Cat. no.:
    46300-494
  • Packaging:
    Bagged
  • Supplier No.:
    845

Specifications

About this item

Microtiter tubes can be autoclaved, sealed and frozen for storage, and they offer additional volume capacity for sample dilution and/or mixing prior to transfer into microtiter plates

  • Autoclavable
  • Manufactured of superior grade polypropylene
  • Can be frozen to -80 °C ( -112 °F)

Ideal for a wide variety of laboratory procedures, such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, blood bank sample freezer storage, and transport of specimens and reagents.