SP Wilmad-LabGlass Soap Film Flowmeters, SP Industries
Supplier: Avantor
Certificates
About this item
Made of borosilicate glass, these soap film bubble flowmeters are used in routine analytical laboratory gas chromatography to measure flow rate of the carrier gas.
A soap bubble, formed in the bottom of the graduated tube, is pushed upward by gas from exit end of the chromatograph. Flow rate is determined by measurement of the volume through which the bubble travels in a defined period of time. Includes LG-4340-100 rubber bulb.
Customers Who Bought This Also Bought