Order Entry
Canada
ContactUsLinkComponent
Anti-NRGN Mouse Monoclonal Antibody [clone: 898502]
Anti-NRGN Mouse Monoclonal Antibody [clone: 898502]
Catalog # 102887-402
Supplier:  R&D Systems
Anti-NRGN Mouse Monoclonal Antibody [clone: 898502]
Catalog # 102887-402
Supplier:  R&D Systems
Supplier Number:  MAB7947

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurogranin
  • Antigen symbol:
    NRGN
  • Clonality:
    Monoclonal
  • Clone:
    898502
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG3
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    64356
  • Antigen synonyms:
    hng|calmodulin-binding protein|NRGN|neurogranin (protein kinase C substrate|neurogranin|protein kinase C substrate|RC3)|RC3
  • Amino acid number:
    Met1 to Asp78
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    E. coli -derived recombinant human Neurogranin Met1-Asp78; Accession # Q92686
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102887-402
  • Supplier No.:
    MAB7947

Specifications

About this item

Type: Primary
Antigen: NRGN
Clonality: Monoclonal
Clone: 898502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat