Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Phospholipase C Gamma 1
- Antigen symbol:PLCG1
- Clonality:Monoclonal
- Clone:1016A
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µL
- Form:Supplied as a 0.2 μm filtered solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:5335
- Antigen synonyms:PLCgamma1|PLC1|PLC148|PLC-148|PLC-II|1-phosphatidyl-D-myo-inositol-4,5-bisphosphate|PLC-gamma-1|inositoltrisphosphohydrolase|PLCG1
- Modification:phospho-Tyr783
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile
- Shipping temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:
Phosphopeptide containing the human/mouse/rat PLC-γ1 (Y783) site.
Accession # P19174
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. no.:102887-126
- Supplier No.:MAB74541
Specifications
About this item
Type: Primary
Antigen Symbol: PLCG1
Antigen Name: phospholipase C, gamma 1
Antigen Synonyms: NCKAP3, PLC-II, PLC1, PLC148, PLCgamma1
Clonality: Monoclonal
Clone: 1016A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat