Order Entry
Canada
ContactUsLinkComponent
Anti-C10ORF54 Mouse Monoclonal Antibody [clone: 730804]
Anti-C10ORF54 Mouse Monoclonal Antibody [clone: 730804]
Catalog # 102886-944
Supplier:  R&D Systems
Anti-C10ORF54 Mouse Monoclonal Antibody [clone: 730804]
Catalog # 102886-944
Supplier:  R&D Systems
Supplier Number:  MAB71261

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromosome 10 open reading frame 54
  • Antigen symbol:
    C10ORF54
  • Clonality:
    Monoclonal
  • Clone:
    730804
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    64115
  • Antigen synonyms:
    B7-H5|GI24|PP2135|DD1alpha|SISP1|B7H5|VISTA
  • Amino acid number:
    Phe33 to Ala194
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H Phe33-Ala194; Accession # Q9H7M9
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102886-944
  • Supplier No.:
    MAB71261

Specifications

About this item

Type: Primary
Antigen: C10ORF54
Clonality: Monoclonal
Clone: 730804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human