Order Entry
Canada
ContactUsLinkComponent
Anti-ESR1 Mouse Monoclonal Antibody [clone: 681104]
Anti-ESR1 Mouse Monoclonal Antibody [clone: 681104]
  102886-092
Anti-ESR1 Mouse Monoclonal Antibody [clone: 681104]
  102886-092
 :  MAB57152

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Estrogen Receptor 1
  • Antigen symbol:
    ESR1
  • Clonality:
    Monoclonal
  • Clone:
    681104
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    2099
  • Antigen synonyms:
    ER|Era|ESRA|NR3A1|ESR|ESTRR
  • Amino acid number:
    Thr2 to Gln116
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    E. coli -derived recombinant human ER alpha/NR3A1 Thr2-Gln116; Accession # P03372
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102886-092
  • Supplier No.:
    MAB57152

 

 

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: 681104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human