Order Entry
Canada
ContactUsLinkComponent
Anti-CADM3 Mouse Monoclonal Antibody [clone: 730014]
Anti-CADM3 Mouse Monoclonal Antibody [clone: 730014]
  102885-614
Anti-CADM3 Mouse Monoclonal Antibody [clone: 730014]
  102885-614
 :  MAB36781

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Cell Adhesion Molecule 3
  • Antigen symbol:
    CADM3
  • Clonality:
    Monoclonal
  • Clone:
    730014
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    57863
  • Antigen synonyms:
    synCAM3|TSLL1|IGSF4B|Necl-1|NECL1|BIgR
  • Amino acid number:
    Pro21 to Tyr329
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human IGSF4B/SynCAM3 Pro21-Tyr329; Accession # Q8N126
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102885-614
  • Supplier No.:
    MAB36781

 

 

Type: Primary
Antigen: CADM3
Clonality: Monoclonal
Clone: 730014
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human