Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Cytokeratin 19
- Antigen symbol:CK19
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:3880
- Antigen synonyms:type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- Amino acid number:Thr2 to Leu400
- Storage temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:E. coli -derived recombinant human Cytokeratin 19 Thr2-Leu400; Accession # P08727
- Purification:Antigen Affinity-purified
- Cat. no.:102884-308
- Supplier No.:IC3506G
Type: Primary
Antigen: CK19
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Sheep
Isotype:
Reactivity: