Order Entry
Canada
ContactUsLinkComponent
Anti-SEMA4D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 758726]
Anti-SEMA4D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 758726]
Catalog # 102883-708
Supplier:  R&D Systems
Anti-SEMA4D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 758726]
Catalog # 102883-708
Supplier:  R&D Systems
Supplier Number:  FAB74701P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Semaphorin 4D
  • Antigen symbol:
    SEMA4D
  • Clonality:
    Monoclonal
  • Clone:
    758726
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    10507
  • Antigen synonyms:
    C9orf164|M-sema-G|SEMAJ|CD100|coll-4
  • Amino acid number:
    Met22 to Arg734
  • Storage temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Chinese hamster ovary cell line CHO-derived recombinant human Semaphorin 4D/CD100 Met22-Arg734; Accession # Q92854
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102883-708
  • Supplier No.:
    FAB74701P

Specifications

About this item

Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 758726
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human