Order Entry
Canada
ContactUsLinkComponent
 
Anti-FLT4 Goat Polyclonal Antibody (PE (Phycoerythrin))
Catalog # 102883-690
Supplier:  R&D Systems
Anti-FLT4 Goat Polyclonal Antibody (PE (Phycoerythrin))
Catalog # 102883-690
Supplier:  R&D Systems
Supplier Number:  FAB743P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FMS Related Tyrosine Kinase 4
  • Antigen symbol:
    FLT4
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    14257
  • Antigen synonyms:
    VEGFR3|LMPH1A|VEGFR-3|VEGF Receptor 3|fms related tyrosine kinase 4|FLT41|FLT-4|Vascular endothelial growth factor receptor 3|PCL
  • Amino acid number:
    Tyr25 to Asp770
  • Storage temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse VEGF R3/Flt-4 Tyr25-Asp770; Accession # P35917
  • Purification:
    Antigen Affinity-purified
  • Cat. no.:
    102883-690
  • Supplier No.:
    FAB743P

Specifications

About this item

Type: Primary
Antigen Symbol: FLT4
Antigen Name: FMS-like tyrosine kinase 4
Antigen Synonyms: AI323512, Chy, Flt-4, VEGFR-3, VEGFR3
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse