Order Entry
Canada
ContactUsLinkComponent
Anti-FSHR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 626717]
Anti-FSHR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 626717]
Catalog # 102883-306
Supplier:  R&D Systems
Anti-FSHR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 626717]
Catalog # 102883-306
Supplier:  R&D Systems
Supplier Number:  FAB65591A

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Follicle Stimulating Hormone Receptor
  • Antigen symbol:
    FSHR
  • Clonality:
    Monoclonal
  • Clone:
    626717
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    2492
  • Antigen synonyms:
    ODG1|FSHR1|LGR1|FSHRO
  • Storage temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    NS0 mouse myeloma cell line transfected with human FSH R; Accession # M65085
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102883-306
  • Supplier No.:
    FAB65591A

Specifications

About this item

Type: Primary
Antigen Symbol: FSHR
Antigen Name: follicle stimulating hormone receptor
Antigen Synonyms: FSHR1, FSHRO, LGR1, ODG1
Clonality: Monoclonal
Clone: 626717
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human