Order Entry
Canada
ContactUsLinkComponent
Anti-ACKR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11G8]
Anti-ACKR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11G8]
  102882-658
Anti-ACKR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11G8]
  102882-658
 :  FAB4227P-025

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Atypical Chemokine Receptor 3
  • Antigen symbol:
    ACKR3
  • Clonality:
    Monoclonal
  • Clone:
    11G8
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Size:
    25 tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    12778
  • Antigen synonyms:
    CXC-R7|RDC-1|CXCR-7|Cxcr7|Rdc1|Cmkor1|AW541270
  • Storage temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Human CXCR7 encoding plasmid; Accession # P25106
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102882-658
  • Supplier No.:
    FAB4227P-025

 

 

Type: Primary
Antigen: ACKR3
Clonality: Monoclonal
Clone: 11G8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse