Order Entry
Canada
ContactUsLinkComponent
Anti-AXL Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 108724]
Anti-AXL Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 108724]
  102881-248
Anti-AXL Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 108724]
  102881-248
 :  FAB154N

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    AXL Receptor Tyrosine Kinase
  • Antigen symbol:
    AXL
  • Clonality:
    Monoclonal
  • Clone:
    108724
  • Conjugation:
    Alexa Fluor® 700
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    558
  • Antigen synonyms:
    JTK11|Tyro7|UFO|ARK
  • Amino acid number:
    Met1 to Pro440
  • Storage temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human Axl Met1-Pro440; Accession # AAA61243
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. no.:
    102881-248
  • Supplier No.:
    FAB154N

 

 

Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 108724
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human