Order Entry
Canada
ContactUsLinkComponent
 
Anti-IL27RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191106]
 
Anti-IL27RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191106]

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Catalog No:
  • 102880-490
  • 102880-492
  • Antigen symbol:
  • IL27RA
  • IL27RA
  • Antigen name:
  • Interleukin 27 Receptor Alpha
  • Interleukin 27 Receptor Alpha
  • Antigen synonyms:
  • WSX1|Interleukin-27 receptor subunit alpha|zcytor1|IL27R|UNQ296/PRO336|CRL1|TCCR|IL-27RA|Cytokine receptor WSX-1|IL-27 receptor subunit alpha
  • WSX1|Interleukin-27 receptor subunit alpha|zcytor1|IL27R|UNQ296/PRO336|CRL1|TCCR|IL-27RA|Cytokine receptor WSX-1|IL-27 receptor subunit alpha
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 191106
  • 191106
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 9466
  • 9466
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Mouse myeloma cell line NS0-derived recombinant human IL-27 R alpha/WSX‑1/TCCR Gly34-Lys516; Accession # Q6UWB1
  • Mouse myeloma cell line NS0-derived recombinant human IL-27 R alpha/WSX‑1/TCCR Gly34-Lys516; Accession # Q6UWB1
  • Amino acid number:
  • Gly34 to Lys516
  • Gly34 to Lys516
  • Form:
  • Supplied in a saline solution containing BSA and Sodium Azide.
  • Supplied in a saline solution containing BSA and Sodium Azide.
  • Purification:
  • Protein A or G purified from hybridoma culture supernatant
  • Protein A or G purified from hybridoma culture supernatant
  • Storage temperature:
  • Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping temperature:
  • The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.

 

Frequently Bought Together