Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Platelet-derived Growth Factor Receptor alpha
- Antigen symbol:PDGFRA
- Clonality:Monoclonal
- Clone:PRa292
- Conjugation:Alexa Fluor® 700
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:5156
- Antigen synonyms:platelet-derived growth factor alpha chain|platelet-derived growth factor A chain|CD140A|platelet-derived growth factor alpha isoform 2 preproprotein|PDGF-1|PDGFR2|platelet-derived growth factor subunit A|RHEPDGFRA,PDGF A-chain|PDGF subunit A
- Storage temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Human osteosarcoma cell membrane extracts
- Purification:Protein A or G purified from ascites
- Cat. no.:102880-304
- Supplier No.:FAB1264N
Specifications
About this item
Type: Primary
Antigen: PDGFRA
Clonality: Monoclonal
Clone: PRa292
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human