ContactUsLinkComponent
Injector Lubricant for Agilent HPLC Systems, Restek
Injector Lubricant for Agilent HPLC Systems, Restek
Catalog # 10057-992
Supplier:  Restek
undefined
Injector Lubricant for Agilent HPLC Systems, Restek
Catalog # 10057-992
Supplier:  Restek
Supplier Number:  26520
Specifications
  • Description:
    Injector Lubricant, PTFE, Similar to Agilent# 79841-65501
  • Size:
    10 g
  • Cat. no.:
    10057-992
  • Supplier No.:
    26520
Specifications
About this item
For use with Agilent 1050 and 1100 HPLC systems.