Orders LinkContactUsLinkComponent
Acrylic Pipe Demonstration
Acrylic Pipe Demonstration
Acrylic Pipe Demonstration
Catalog # 470219-530
Supplier:   Wards
Supplier #:  PH100LCB
Acrylic Pipe Demonstration
Catalog # 470219-530
Supplier:  Wards
Acrylic Pipe Demonstration
Acrylic Pipe Demonstration
Acrylic Pipe Demonstration
Price
34,25 $
Log in for availability
Each
Product Details & Documents
Show students how light reflects inside an acrylic pipe.
  • Transmits light like fiber optic cable
This hard acrylic 1/2" diameter light pipe is 2 feet of pipe coiled up at one end to demonstrate light transmission end to end. This light pipe is approx. 9" wide due to the coil in it.
Specifications
  • Description:
    Acrylic Pipe Demonstration