Spring Large - Compression
Spring Large - Compression
Catalog # 470219-412
Supplier: Wards
Supplier #: PH0115C
Jump to:
Product Details & Documents
Excellent demonstration of compression for hooke's law. Approximately 300mm long with a 10mm diameter and wire of 19 SWG.
Specifications
- Description:Spring Large - Compression