Orders LinkContactUsLinkComponent
Vernier® Motion Encoder Cart
Vernier® Motion Encoder Cart
New Product
Vernier® Motion Encoder Cart
Vernier® Motion Encoder Cart
The Motion Encoder Cart is a dynamics cart compatible with the Vernier Dynamics Cart and Track System. The Motion Encoder Cart replaces a motion detector when used for dynamics cart-on-track activities. An optical sensor beneath the dynamics cart senses the passage of the cart over a marked strip on the track. The position information is sent as an encoded IR signal to a receiver at the track’s end. By avoiding the possibility of stray sound reflections, this optical-only system provides excellent, repeatable, and noise-resistant motion data.
The cart must be used with the Motion Encoder Receiver, and the track must have the Motion Encoder Track Strip installed.
Product Family Options
  • Description
  • Vernier motion encoder cart
    Each
    Each - $393.09
    $393.09
    Each
       $393.09
     : DTS-CART-MEC
    New Product
    New Product
  • Vernier motion encoder cart with reciever and track strip
    Each
    Each - $555.09
    $555.09
    Each
       $555.09
     : DTS-MEC
    New Product
    New Product
  • Motion encoder track strip, 2.2 m
    Each
    Each - $9.09
    $9.09
    Each
       $9.09
     : METS-LONG
    New Product
    New Product
  • Vernier motion encoder receiver, BTD
    Each
    Each - $160.09
    $160.09
    Each
       $160.09
     : MEC-BTD
    New Product
    New Product
Accessories
  •  
    Description
  • CA470303-824
    125 g masses for Vernier dynamics carts, pack of 4
    Each - $29.09