Orders LinkContactUsLinkComponent
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
Certificates
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
6 Options Available Below
Product Details & Documents
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Product Family Options
Product Information
  • Material
    Septum
    Cap Size
    Cap color
    Cap type
    Availability
    Price
  • PP
    Bonded Silicone/PTFE, low durometer
    9-425 mm
    Blue
    Screw cap, knurled
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $79.93
    $79.93
    Pack of 100
       $79.93
    Minimum order quantity is 1
    Supplier Number: 9502S-10C-B
  • PP
    Bonded Silicone/PTFE, low durometer
    9-425 mm
    Blue
    Screw cap, knurled
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $755.13
    $755.13
    Case of 1,000
       $755.13
    Minimum order quantity is 1
    Supplier Number: 9502S-10C-B-M
  • PP
    Bonded Silicone/PTFE, pre-slit
    9-425 mm
    Blue
    Screw cap, knurled
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $99.16
    $99.16
    Pack of 100
       $99.16
    Minimum order quantity is 1
    Supplier Number: 9502S-30C-B
  • PP
    Bonded Silicone/PTFE, pre-slit
    9-425 mm
    Blue
    Screw cap, knurled
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $928.84
    $928.84
    Case of 1,000
       $928.84
    Minimum order quantity is 1
    Supplier Number: 9502S-30C-B-M
  • PP
    Bonded Silicone/PTFE
    9-425 mm
    Blue
    Screw cap, knurled
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $135.81
    $135.81
    Pack of 100
       $135.81
    Supplier Number: 9502S-16B
  • PP
    Fitted Silicone/PTFE
    9-425 mm
    Blue
    Screw cap, smooth sided, Robo Style
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $635.18
    $635.18
    Case of 1,000
       $635.18
    Supplier Number: 9502S-05-B-M