Order Entry
Canada
ContactUsLinkComponent
Bipolar Cushing Forceps, OR Grade, Sklar
Bipolar Cushing Forceps, OR Grade, Sklar
Catalog # 10184-544
Supplier:  Sklar
undefined
Bipolar Cushing Forceps, OR Grade, Sklar
Catalog # 10184-544
Supplier:  Sklar
Supplier Number:  91-6102

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    152 mm (6")
  • Material:
    Stainless Steel with Non-Conductive Nylon Coating
  • Cat. no.:
    10184-544
  • Supplier No.:
    91-6102

Specifications

About this item

Bipolar Cushing Forceps are industry standard.